Gene Transcripts

Gene Symbol
RMND1
Entrez Gene ID
55005
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3
Chromosome Location
Chr. 6: 151404548 - 151452181 on Build GRCh38
UniGene ID
Hs.486835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271937.1 NP_001258866.1 5 441
NM_017909.3 NP_060379.2 6 959
XM_005267040.3 XP_005267097.1 5 440
XM_017010988.1 XP_016866477.1 6 958
GenBank mRNA AK000634.1 - 6 902
AK026480.1 - 5 461
AK292339.1 - 6 934
BC106065.1 - 6 921 6165385
BC119683.1 - 6 824 40116343
BG547724.1 - 5 380

View Full Product Details