Gene Transcripts
- Gene Symbol
- RNASET2
- Entrez Gene ID
- 8635
- Gene Name
- ribonuclease T2
- Gene Aliases
- RNASE6PL, bA514O12.3
- Chromosome Location
- Chr. 6: 166929509 - 166956589 on Build GRCh38
- UniGene ID
- Hs.529989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003730.4 |
NP_003721.2
|
4, 5 |
676 |
|
XM_017011397.1 |
XP_016866886.1
|
5, 6 |
455 |
|
XM_017011398.1 |
XP_016866887.1
|
3, 4 |
577 |
|
XM_017011399.1 |
XP_016866888.1
|
4, 5 |
637 |
|
GenBank mRNA |
AJ419865.1 |
- |
4, 5 |
630 |
|
AK315467.1 |
- |
4, 5 |
637 |
|
BC001660.2 |
- |
4, 5 |
602 |
2959583 |
BC001819.2 |
- |
4, 5 |
602 |
2959583 |
BC036282.1 |
- |
4, 5 |
663 |
|
BC039713.1 |
- |
4, 5 |
605 |
5225327 |
BC051912.1 |
- |
4, 5 |
615 |
5227463 |
U85625.2 |
- |
4, 5 |
633 |
|
View Full Product Details