Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286277.1 | NP_001273206.1 | 10 | 1239 | |
NM_001286278.1 | NP_001273207.1 | 10 | 1269 | ||
NM_001286279.1 | NP_001273208.1 | 10 | 1269 | ||
NM_006322.5 | NP_006313.1 | 10 | 1269 | ||
XM_005268293.1 | XP_005268350.1 | 10 | 1264 | ||
XM_011537463.2 | XP_011535765.1 | 11 | 1198 | ||
XM_011537464.2 | XP_011535766.1 | 10 | 2002 | ||
XM_017020323.1 | XP_016875812.1 | 11 | 1616 | ||
GenBank mRNA | AF042378.1 | - | 10 | 1148 | |
AJ003061.1 | - | 10 | 1064 | ||
AJ003062.1 | - | 10 | 1064 | ||
AK302537.1 | - | 10 | 1186 | ||
AK314785.1 | - | 10 | 1269 | ||
BC007763.2 | - | 10 | 1219 | ||
BC013781.1 | - | 10 | 1209 | 3867019 | |
BC046634.2 | - | 10 | 1252 | 5295831 | |
BP381545.1 | - | 3 | 144 | ||
BT009772.1 | - | 10 | 1064 |