Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001313972.1 | NP_001300901.1 | 7 | 1600 | |
NM_006472.5 | NP_006463.3 | 8 | 1969 | ||
XM_017000085.1 | XP_016855574.1 | 7 | 2047 | ||
GenBank mRNA | AK226090.1 | - | 8 | 1881 | |
AK301203.1 | - | 4 | 1089 | ||
BC024305.1 | - | 8 | 1499 | ||
BX404012.2 | - | 1 | 98 | ||
BX537824.1 | - | 5 | 1351 | ||
S73591.1 | - | 8 | 1825 |