Gene Transcripts

Gene Symbol
NPM2
Entrez Gene ID
10361
Gene Name
nucleophosmin/nucleoplasmin 2
Gene Aliases
Chromosome Location
Chr. 8: 22023308 - 22036908 on Build GRCh38
UniGene ID
Hs.131055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286680.1 NP_001273609.1 7 1133
NM_182795.1 NP_877724.1 6 719
XM_011544360.2 XP_011542662.1 5 1355
XM_011544362.2 XP_011542664.1 7 752
XM_011544363.2 XP_011542665.1 6 633
XM_011544364.2 XP_011542666.1 6 1042
XM_017012947.1 XP_016868436.1 4 1245
XM_017012948.1 XP_016868437.1 7 1352
XM_017012949.1 XP_016868438.1 6 1518
XM_017012950.1 XP_016868439.1 6 1226
GenBank mRNA AK094267.1 - 6 1494
AY262113.1 - 7 809
AY262114.1 - 6 719

View Full Product Details