Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100816.1 | NP_001094286.1 | 2 | 1204 | |
NM_004270.4 | NP_004261.1 | 2 | 920 | ||
GenBank mRNA | AK308393.1 | - | 2 | 884 | |
BC005250.1 | - | 2 | 874 | ||
BC006968.1 | - | 2 | 874 |