Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004784.2 | NP_004775.1 | 14 | 2933 | |
XM_006714416.3 | XP_006714479.1 | 14 | 2989 | ||
XM_017008839.1 | XP_016864328.1 | 14 | 2896 | ||
XM_017008840.1 | XP_016864329.1 | 14 | 2766 | ||
XM_017008841.1 | XP_016864330.1 | 13 | 4607 | ||
XM_017008842.1 | XP_016864331.1 | 13 | 2216 | ||
XM_017008843.1 | XP_016864332.1 | 11 | 1389 | ||
XM_017008844.1 | XP_016864333.1 | 13 | 1646 | ||
GenBank mRNA | AF074924.1 | - | 14 | 2933 | |
AF076605.1 | - | 13 | 2719 | ||
BC109309.1 | - | 14 | 2780 | 40035856 | |
BC109310.1 | - | 14 | 2780 | 40035859 |