Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr. 15: 89681531 - 89690784 on Build GRCh38
UniGene ID
Hs.31034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271572.1 NP_001258501.1 2 280
NM_003847.2 NP_003838.1 2 280
GenBank mRNA AB015594.1 - 2 223
AF093668.1 - 2 185
AK001415.1 - 2 273
AK300978.1 - 2 158
AK313318.1 - 2 194
BC009697.1 - 2 174 3878406
BC101164.2 - 2 134 40015908
BC114384.1 - 2 134 40004153
CR542046.1 - 2 128
CR542075.1 - 2 128

View Full Product Details