Gene Transcripts

Gene Symbol
LIPF
Entrez Gene ID
8513
Gene Name
lipase F, gastric type
Gene Aliases
GL, HGL, HLAL
Chromosome Location
Chr. 10: 88664389 - 88678815 on Build GRCh38
UniGene ID
Hs.523130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198828.1 NP_001185757.1 6 604
NM_001198829.1 NP_001185758.1 7 784
NM_001198830.1 NP_001185759.1 7 685
NM_004190.3 NP_004181.1 6 703
XM_011540311.1 XP_011538613.1 6 645
GenBank mRNA A01046.1 - 6 635
AK223468.1 - 6 648
AK301310.1 - 6 553
AK301320.1 - 7 732
AK308224.1 - 6 651
AK312940.1 - 6 650
AL833751.1 - 7 616
BC112272.1 - 5 622 8327740
BC113711.1 - 5 611 8322763
X05997.1 - 6 633

View Full Product Details