Gene Transcripts

Gene Symbol
TUFT1
Entrez Gene ID
7286
Gene Name
tuftelin 1
Gene Aliases
Chromosome Location
Chr. 1: 151540294 - 151583583 on Build GRCh38
UniGene ID
Hs.489922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126337.1 NP_001119809.1 4 364
NM_001301317.1 NP_001288246.1 6 574
NM_020127.2 NP_064512.1 5 439
XM_011509961.2 XP_011508263.1 3 273
XM_017002223.1 XP_016857712.1 5 515
XM_017002224.1 XP_016857713.1 4 636
GenBank mRNA AF254260.1 - 5 439
AK075001.1 - 4 361
AK223161.1 - 5 435
AK298066.1 - 6 574
AK299263.1 - 4 362
AK299925.1 - 4 352
AK315414.1 - 5 435
AL136917.1 - 5 427
AY358610.1 - 4 358
BC002933.1 - 4 332 3956873
BC008301.1 - 5 419 3506004

View Full Product Details