Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018412.3 | NP_060882.2 | 5 | 688 | |
NM_021908.2 | NP_068708.1 | 5 | 688 | ||
GenBank mRNA | AF234882.1 | - | 5 | 687 | |
AF234883.1 | - | 5 | 687 | ||
AF415174.1 | - | 3 | 115 | ||
AJ277291.1 | - | 4 | 323 | ||
AK093212.1 | - | 6 | 937 | ||
AK292372.1 | - | 5 | 514 | ||
AK297510.1 | - | 5 | 565 | ||
AK297861.1 | - | 5 | 396 | ||
AK298540.1 | - | 5 | 510 | ||
AY009152.1 | - | 5 | 679 | ||
AY009153.1 | - | 5 | 679 | ||
BC030954.1 | - | 6 | 493 | ||
BC075855.1 | - | 5 | 688 | 5562032 | |
KJ534947.1 | - | 5 | 474 |