Gene Transcripts

Gene Symbol
LEMD3
Entrez Gene ID
23592
Gene Name
LEM domain containing 3
Gene Aliases
MAN1
Chromosome Location
Chr. 12: 65169571 - 65248361 on Build GRCh38
UniGene ID
Hs.744150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167614.1 NP_001161086.1 1 222
NM_014319.4 NP_055134.2 1 222
GenBank mRNA AF112299.2 - 1 202
AK304317.1 - 1 210
AK312473.1 - 1 222

View Full Product Details