Gene Transcripts

Gene Symbol
MED31
Entrez Gene ID
51003
Gene Name
mediator complex subunit 31
Gene Aliases
3110004H13Rik, CGI-125, Soh1
Chromosome Location
Chr. 17: 6643313 - 6651804 on Build GRCh38
UniGene ID
Hs.567493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016060.2 NP_057144.1 4 980
GenBank mRNA AF151883.1 - 4 952

View Full Product Details