Gene Transcripts

Gene Symbol
RPTOR
Entrez Gene ID
57521
Gene Name
regulatory associated protein of MTOR complex 1
Gene Aliases
KOG1, Mip1
Chromosome Location
Chr. 17: 80544825 - 80966373 on Build GRCh38
UniGene ID
Hs.133044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163034.1 NP_001156506.1 22 3381
NM_020761.2 NP_065812.1 26 3855
GenBank mRNA AB037724.1 - 22 2537
AB082951.1 - 26 3050
AY090663.1 - 26 3412
BC033258.1 - 2 70
BC136652.1 - 26 3245 9020642
BC136654.1 - 26 3245 9020644

View Full Product Details