Gene Transcripts
- Gene Symbol
- BPHL
- Entrez Gene ID
- 670
- Gene Name
- biphenyl hydrolase-like (serine hydrolase)
- Gene Aliases
- BPH-RP, MCNAA, VACVASE
- Chromosome Location
- Chr. 6: 3118376 - 3153198 on Build GRCh38
- UniGene ID
- Hs.10136
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001302777.1 |
NP_001289706.1
|
7, 8 |
1382 |
|
NM_004332.3 |
NP_004323.2
|
6, 7 |
1154 |
|
NR_026648.2 |
|
8, 9 |
1450 |
|
NR_026649.2 |
|
7, 8 |
1222 |
|
GenBank mRNA |
AJ617684.1 |
- |
6, 7 |
802 |
|
AK297518.1 |
- |
7, 8 |
906 |
|
AK314909.1 |
- |
7, 8 |
1036 |
|
AL832502.1 |
- |
6, 7 |
3149 |
|
BC037778.1 |
- |
8, 9 |
1455 |
4799692 |
BC106901.1 |
- |
6, 7 |
869 |
40031474 |
X81372.1 |
- |
7, 8 |
950 |
|
View Full Product Details