Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002257.2 | NP_001002257.1 | 3 | 422 | |
NM_001304445.1 | NP_001291374.1 | 5 | 827 | ||
NM_001304446.1 | NP_001291375.1 | 2 | 274 | ||
NM_182551.4 | NP_872357.2 | 4 | 614 | ||
XM_005264244.1 | XP_005264301.1 | 4 | 576 | ||
XM_005264245.3 | XP_005264302.1 | 3 | 428 | ||
XM_011532741.2 | XP_011531043.1 | 4 | 565 | ||
XM_011532742.2 | XP_011531044.1 | 4 | 392 | ||
XM_011532743.2 | XP_011531045.1 | 3 | 379 | ||
XM_011532744.2 | XP_011531046.1 | 3 | 417 | ||
XM_011532745.1 | XP_011531047.1 | 4 | 576 | ||
XM_017003746.1 | XP_016859235.1 | 4 | 459 | ||
XM_017003747.1 | XP_016859236.1 | 4 | 576 | ||
GenBank mRNA | AK095284.1 | - | 4 | 579 | |
AK296965.1 | - | 3 | 407 | ||
AK297262.1 | - | 2 | 254 | ||
AK302562.1 | - | 5 | 780 | ||
AY358702.1 | - | 4 | 583 | ||
BC146817.1 | - | 3 | 412 | 8860419 | |
BP872689.1 | - | 3 | 422 | ||
BX647953.1 | - | 3 | 378 |