Gene Transcripts
- Gene Symbol
- PLEKHG2
- Entrez Gene ID
- 64857
- Gene Name
- pleckstrin homology and RhoGEF domain containing G2
- Gene Aliases
- ARHGEF42, CLG, LDAMD
- Chromosome Location
- Chr. 19: 39412582 - 39428415 on Build GRCh38
- UniGene ID
- Hs.631574
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022835.2 |
NP_073746.2
|
16, 17 |
1824 |
|
XM_005259163.2 |
XP_005259220.1
|
16, 17 |
1645 |
|
XM_006723334.2 |
XP_006723397.1
|
16, 17 |
2175 |
|
XM_011527232.2 |
XP_011525534.1
|
16, 17 |
1751 |
|
XM_017027150.1 |
XP_016882639.1
|
15, 16 |
2316 |
|
XM_017027151.1 |
XP_016882640.1
|
15, 16 |
2307 |
|
GenBank mRNA |
AK024429.1 |
- |
16, 17 |
1824 |
|
AK026111.1 |
- |
16, 17 |
1633 |
|
AK302476.1 |
- |
16, 17 |
1448 |
|
BC013426.1 |
- |
6, 7 |
315 |
|
BC075858.1 |
- |
12, 13 |
1102 |
|
View Full Product Details