Gene Transcripts

Gene Symbol
SGMS1
Entrez Gene ID
259230
Gene Name
sphingomyelin synthase 1
Gene Aliases
MOB, MOB1, SMS1, TMEM23, hmob33
Chromosome Location
Chr. 10: 50305585 - 50623977 on Build GRCh38
UniGene ID
Hs.654698
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147156.3 NP_671512.1 11 3684
XM_005269675.1 XP_005269732.1 6 3376
XM_011539582.2 XP_011537884.1 8 3288
XM_011539583.2 XP_011537885.1 6 3165
GenBank mRNA AY280959.1 - 11 3650
BC042899.1 - 5 2945 4342258
Y14155.1 - 1 1517

View Full Product Details