Gene Transcripts
- Gene Symbol
- RP9
- Entrez Gene ID
- 6100
- Gene Name
- retinitis pigmentosa 9 (autosomal dominant)
- Gene Aliases
- PAP-1, PAP1
- Chromosome Location
- Chr. 7: 33094797 - 33109390 on Build GRCh38
- UniGene ID
- Hs.326805
- Species
- Homo sapiens
- Gene Symbol
- RP9P
- Entrez Gene ID
- 441212
- Gene Name
- retinitis pigmentosa 9 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 32916815 - 32943170 on Build GRCh38
- UniGene ID
- Hs.648086
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003500.1 |
|
6 |
584 |
|
GenBank mRNA |
AF274938.1 |
- |
5 |
294 |
|
BC050570.1 |
- |
7 |
689 |
|
View Full Product Details