Gene Transcripts

Gene Symbol
RP9
Entrez Gene ID
6100
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Gene Aliases
PAP-1, PAP1
Chromosome Location
Chr. 7: 33094797 - 33109390 on Build GRCh38
UniGene ID
Hs.326805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203288.1 NP_976033.1 5 474
XM_011515468.2 XP_011513770.1 5 950
GenBank mRNA BC025928.1 - 5 474

View Full Product Details