Gene Transcripts

Gene Symbol
TMEM88
Entrez Gene ID
92162
Gene Name
transmembrane protein 88
Gene Aliases
Chromosome Location
Chr. 17: 7854886 - 7856099 on Build GRCh38
UniGene ID
Hs.389669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203411.1 NP_981956.1 2 501
XM_005256856.3 XP_005256913.1 2 723
GenBank mRNA AK000032.1 - 2 502
BC057812.1 - 2 501 30332216

View Full Product Details