Gene Transcripts

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
Chromosome Location
Chr. 4: 186191520 - 186213463 on Build GRCh38
UniGene ID
Hs.587231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK122600.1 - 6 817
AK126473.1 - 7 751
AY422002.1 - 6 1082
BC060857.1 - 6 948 30333559

View Full Product Details