Gene Transcripts

Gene Symbol
Ptx3
Entrez Gene ID
19288
Gene Name
pentraxin related gene
Gene Aliases
AI607804, TSG-14
Chromosome Location
Chr. 3: 66219887 - 66225806 on Build GRCm38
UniGene ID
Mm.276776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008987.3 NP_033013.3 1 255
GenBank mRNA BC022176.1 - 1 255
BQ442682.1 - 1 142
CA529293.1 - 1 255
X83601.1 - 1 179

View Full Product Details