Gene Transcripts

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
Chromosome Location
Chr. 1: 198156978 - 198322420 on Build GRCh38
UniGene ID
Hs.24119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133494.2 NP_598001.1 6 727
XM_017000344.1 XP_016855833.1 6 729
XM_017000345.1 XP_016855834.1 7 836
XM_017000346.1 XP_016855835.1 6 729
XM_017000347.1 XP_016855836.1 6 729
XM_017000348.1 XP_016855837.1 6 729
GenBank mRNA AB062450.1 - 5 446
AK313411.1 - 6 527

View Full Product Details