Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
20431
Gene Name
premelanosome protein
Gene Aliases
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87
Chromosome Location
Chr. 10: 128705168 - 128720238 on Build GRCm38
UniGene ID
Mm.16756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021882.4 NP_068682.2 3 318
GenBank mRNA AK012808.1 - 3 319
AK131956.1 - 3 320
BC075667.1 - 3 305
BC082555.1 - 3 304 30541702
U14133.1 - 3 293

View Full Product Details