Gene Transcripts

Gene Symbol
Nxt2
Entrez Gene ID
237082
Gene Name
nuclear transport factor 2-like export factor 2
Gene Aliases
6330587F24Rik, AI787442, P15-2
Chromosome Location
Chr. X: 142226770 - 142239700 on Build GRCm38
UniGene ID
Mm.41899
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161430.2 NP_001154902.1 5 879
NM_001290532.1 NP_001277461.1 5 964
NM_001290533.1 NP_001277462.1 4 970
NM_172782.4 NP_766370.2 4 916
GenBank mRNA AK032102.1 - 4 887
BC064727.1 - 4 891
BC068166.1 - 4 869 30361776
BC094570.1 - 4 833 30361291
BC118503.1 - 4 844 40106658

View Full Product Details