Gene Transcripts

Gene Symbol
Mesp2
Entrez Gene ID
17293
Gene Name
mesoderm posterior 2
Gene Aliases
bHLHc6
Chromosome Location
Chr. 7: 79810727 - 79813431 on Build GRCm38
UniGene ID
Mm.31472
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008589.2 NP_032615.2 2 1573
GenBank mRNA AK146740.1 - 2 1053
U71125.1 - 2 1577

View Full Product Details