Gene Transcripts

Gene Symbol
Mtnr1a
Entrez Gene ID
17773
Gene Name
melatonin receptor 1A
Gene Aliases
MR, MelR
Chromosome Location
Chr. 8: 45069210 - 45089233 on Build GRCm38
UniGene ID
Mm.5133
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008639.2 NP_032665.1 2 890
GenBank mRNA BC116900.1 - 2 963
BC119313.1 - 2 963 8734062
U52222.1 - 2 890

View Full Product Details