Gene Transcripts

Gene Symbol
Nat3
Entrez Gene ID
17962
Gene Name
N-acetyltransferase 3
Gene Aliases
Chromosome Location
Chr. 8: 67523854 - 67548627 on Build GRCm38
UniGene ID
Mm.57193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008674.2 NP_032700.1 3 876
GenBank mRNA AK077364.1 - 3 876

View Full Product Details