Gene Transcripts

Gene Symbol
Obp1a
Entrez Gene ID
18249
Gene Name
odorant binding protein IA
Gene Aliases
OBP-1a, OBP5
Chromosome Location
Chr. X: 78085505 - 78091374 on Build GRCm38
UniGene ID
Mm.430771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008754.2 NP_032780.2 7 577
GenBank mRNA AK017455.1 - 7 578
BC125514.1 - 7 564 40130029
BC125516.1 - 7 564 40130031

View Full Product Details