Gene Transcripts

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
Chromosome Location
Chr. 1: 27355179 - 27368037 on Build GRCh38
UniGene ID
Hs.194694
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297609.1 NP_001284538.1 4, 5 1117
NM_004672.4 NP_004663.3 5, 6 1141
XM_017002771.1 XP_016858260.1 3, 4 510
XM_017002772.1 XP_016858261.1 5, 6 2312
GenBank mRNA AB167411.1 - 5, 6 872
AB208805.1 - 4, 5 586
AF100318.1 - 2, 3 286
BC015914.1 - 1, 2 43

View Full Product Details