Gene Transcripts

Gene Symbol
Fabp5
Entrez Gene ID
16592
Gene Name
fatty acid binding protein 5, epidermal
Gene Aliases
E-FABP, Fabpe, Klbp, PA-FABP, mal1
Chromosome Location
Chr. 3: 10012585 - 10016610 on Build GRCm38
UniGene ID
Mm.741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272097.1 NP_001259026.1 2 183
NM_010634.3 NP_034764.1 2 183
GenBank mRNA AK008782.1 - 2 177
AK011551.1 - 2 179
AK166506.1 - 2 178
AK167389.1 - 2 178
BC002008.1 - 2 163 3490535
BC053042.1 - 1 2415
BC100543.1 - 2 158 30136900
X70100.1 - 2 162

View Full Product Details