Gene Transcripts

Gene Symbol
Olfr30
Entrez Gene ID
18329
Gene Name
olfactory receptor 30
Gene Aliases
MOR2811, MTPCR07
Chromosome Location
Chr. 11: 58454915 - 58455980 on Build GRCm38
UniGene ID
Mm.222836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146878.2 NP_667089.1 1 624
GenBank mRNA AY043304.1 - 1 208
BC119375.1 - 1 624 8734124
X89680.1 - 1 214

View Full Product Details