Gene Transcripts

Gene Symbol
MED13
Entrez Gene ID
9969
Gene Name
mediator complex subunit 13
Gene Aliases
ARC250, DRIP250, HSPC221, THRAP1, TRAP240
Chromosome Location
Chr. 17: 61942605 - 62065983 on Build GRCh38
UniGene ID
Hs.282678
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005121.2 NP_005112.2 16 3500
XM_011525551.2 XP_011523853.1 16 4201
XM_011525552.2 XP_011523854.1 16 4201
XM_011525553.2 XP_011523855.1 13 2915
GenBank mRNA AB011165.2 - 13 2918
AF117754.1 - 17 3500
BC140891.1 - 16 3474 9021759

View Full Product Details