Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027290.3 | NP_081566.2 | 9 | 1417 | |
GenBank mRNA | AK030780.1 | - | 9 | 1362 | |
AK041749.1 | - | 3 | 423 | ||
BC023236.1 | - | 1 | 279 | ||
BC064314.1 | - | 9 | 1780 | ||
BC094576.1 | - | 9 | 1417 | 6853824 | |
BC120687.1 | - | 8 | 1326 | 40129610 | |
BC120689.1 | - | 8 | 1326 | 40129612 |