Gene Transcripts

Gene Symbol
LPA
Entrez Gene ID
4018
Gene Name
lipoprotein(a)
Gene Aliases
AK38, APOA, LP
Chromosome Location
Chr. 6: 160531483 - 160666375 on Build GRCh38
UniGene ID
Hs.520120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005577.2 NP_005568.2 5 548
GenBank mRNA X06290.1 - 473

View Full Product Details