Gene Transcripts

Gene Symbol
Rhot1
Entrez Gene ID
59040
Gene Name
ras homolog gene family, member T1
Gene Aliases
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1
Chromosome Location
Chr. 11: 80209005 - 80267907 on Build GRCm38
UniGene ID
Mm.489674
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163354.1 NP_001156826.1 7 629
NM_001163355.1 NP_001156827.1 7 629
NM_021536.7 NP_067511.4 7 629
GenBank mRNA AK019059.1 - 7 589
AK028597.1 - 7 572
AK031139.1 - 5 291
AK032096.1 - 7 602
AK041119.1 - 7 565
AK053263.1 - 6 486
AK054472.1 - 7 600
AK078955.1 - 7 572
AK138830.1 - 7 570
AK141621.1 - 7 456
AK147749.1 - 7 608
AK147801.1 - 7 634
AK167502.1 - 7 629
AK169811.1 - 7 598
AK191854.1 - 5 263
BC046785.1 - 7 612 6416127
BC058350.1 - 7 544 6815832

View Full Product Details