Gene Transcripts
- Gene Symbol
- LPA
- Entrez Gene ID
- 4018
- Gene Name
- lipoprotein(a)
- Gene Aliases
- AK38, APOA, LP
- Chromosome Location
- Chr. 6: 160531483 - 160666375 on Build GRCh38
- UniGene ID
- Hs.520120
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005577.2 |
NP_005568.2
|
23 |
3733 |
|
GenBank mRNA |
X06290.1 |
- |
|
11182 |
|
View Full Product Details