Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009059.2 | NP_033085.2 | 9 | 1348 | |
GenBank mRNA | AK144440.1 | - | 9 | 1350 | |
AK204851.1 | - | 2 | 89 | ||
AK215408.1 | - | 2 | 134 | ||
BC068121.1 | - | 9 | 1348 | 6415928 | |
KJ650210.1 | - | 8 | 1156 | ||
U54639.1 | - | 9 | 1277 |