Gene Transcripts

Gene Symbol
Myl6
Entrez Gene ID
17904
Gene Name
myosin, light polypeptide 6, alkali, smooth muscle and non-muscle
Gene Aliases
ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3SM, MLC3nm, Myln
Chromosome Location
Chr. 10: 128490861 - 128493896 on Build GRCm38
UniGene ID
Mm.337074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010860.3 NP_034990.1 4 271
GenBank mRNA AK143885.1 - 4 301
AK159044.1 - 1 1581
AK167106.1 - 4 301
AK167191.1 - 4 307
AK168450.1 - 4 322
AK168869.1 - 4 303
BC026760.1 - 4 271 3990090
BC081470.1 - 4 288 6824167
BC089485.1 - 4 294 30277988
BC092214.1 - 4 279 5684607
U04443.1 - 2 227

View Full Product Details