Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025513.3 | NP_079789.1 | 1, 2 | 338 | |
GenBank mRNA | AK006281.1 | - | 1, 2 | 234 | |
AK012689.1 | - | 1, 2 | 339 | ||
BC023669.1 | - | 1, 2 | 333 | 5012335 | |
BC054085.1 | - | 1, 2 | 343 | 3371550 |