Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270439.1 | NP_001257368.1 | 3 | 525 | |
NM_005717.3 | NP_005708.1 | 3 | 516 | ||
GenBank mRNA | AF006088.1 | - | 3 | 258 | |
AF017807.1 | - | 3 | 237 | ||
AK095424.1 | - | 2 | 896 | ||
AK129598.1 | - | 3 | 301 | ||
BC057237.1 | - | 3 | 345 | 3931697 | |
BI758106.1 | - | 3 | 425 | ||
CN291383.1 | - | 3 | 516 |