Gene Transcripts

Gene Symbol
Map6d1
Entrez Gene ID
208158
Gene Name
MAP6 domain containing 1
Gene Aliases
A330066I15Rik
Chromosome Location
Chr. 16: 20233309 - 20241358 on Build GRCm38
UniGene ID
Mm.326383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198599.2 NP_941001.2 3 1220
GenBank mRNA AK079603.1 - 3 1186
AK163656.1 - 3 1184

View Full Product Details