Gene Transcripts

Gene Symbol
Mbip
Entrez Gene ID
217588
Gene Name
MAP3K12 binding inhibitory protein 1
Gene Aliases
4933408E06Rik
Chromosome Location
Chr. 12: 56328306 - 56345894 on Build GRCm38
UniGene ID
Mm.25656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145442.2 NP_663417.1 4 580
GenBank mRNA AK077125.1 - 4 501
AK154318.1 - 4 509
AK171164.1 - 4 509
BC002277.1 - 4 566 3496688

View Full Product Details