Gene Transcripts

Gene Symbol
Sel1l3
Entrez Gene ID
231238
Gene Name
sel-1 suppressor of lin-12-like 3 (C. elegans)
Gene Aliases
2310045A20Rik, AI429585, mKIAA0746
Chromosome Location
Chr. 5: 53107083 - 53213452 on Build GRCm38
UniGene ID
Mm.235020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172710.3 NP_766298.2 7 1406
GenBank mRNA AK030487.1 - 7 1271
AK037220.1 - 7 1270
AK122362.1 - 7 1406
BC094607.1 - 6 951 6837048
BC117915.1 - 6 907 40097358
BC117916.1 - 6 909 40097359

View Full Product Details