Gene Transcripts

Gene Symbol
Tmem60
Entrez Gene ID
212090
Gene Name
transmembrane protein 60
Gene Aliases
BC027828
Chromosome Location
Chr. 5: 20882453 - 20886870 on Build GRCm38
UniGene ID
Mm.347525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177601.3 NP_808269.1 2 753
GenBank mRNA BC027828.1 - 1 530

View Full Product Details