Gene Transcripts

Gene Symbol
Nox3
Entrez Gene ID
224480
Gene Name
NADPH oxidase 3
Gene Aliases
GP91-3, het, nmf250
Chromosome Location
Chr. 17: 3635240 - 3696261 on Build GRCm38
UniGene ID
Mm.328756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198958.2 NP_945196.2 11 1486
GenBank mRNA AY182377.1 - 11 1486
AY573240.1 - 11 1426
BC106862.2 - 11 1486 40047310

View Full Product Details