Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145013.1 | NP_001138485.1 | 3 | 431 | |
NM_138954.3 | NP_620404.1 | 3 | 663 | ||
GenBank mRNA | AK136022.1 | - | 3 | 432 | |
AY070253.1 | - | 3 | 653 | ||
BC069834.1 | - | 3 | 417 | 30474534 |