Gene Transcripts

Gene Symbol
NDRG1
Entrez Gene ID
10397
Gene Name
N-myc downstream regulated 1
Gene Aliases
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5
Chromosome Location
Chr. 8: 133237171 - 133297587 on Build GRCh38
UniGene ID
Hs.372914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135242.1 NP_001128714.1 4 699
NM_001258432.1 NP_001245361.1 2 185
NM_006096.3 NP_006087.2 4 302
XM_011516791.1 XP_011515093.1 4 585
GenBank mRNA AF004162.1 - 3 141
AK095649.1 - 4 251
AK096562.1 - 4 251
AK126924.1 - 4 981
AK223542.1 - 4 204
AK296794.1 - 2 133
AK297342.1 - 251
AK298236.1 - 4 252
AK307829.1 - 4 251
AK308117.1 - 2 134
AU141826.1 - 4 254
AY277597.1 - 3 130
BC003175.1 - 4 230 2957847
CR456842.1 - 3 113
D87953.1 - 4 235
DC353531.1 - 4 302
X92845.1 - 4 223

View Full Product Details