Gene Transcripts

Gene Symbol
Mlec
Entrez Gene ID
109154
Gene Name
malectin
Gene Aliases
2410014A08Rik, AI850530, ESTM19, X83352, mKIAA0152
Chromosome Location
Chr. 5: 115142981 - 115158176 on Build GRCm38
UniGene ID
Mm.153963
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175403.3 NP_780612.2 5 1843
GenBank mRNA AK031525.1 - 5 1817
AK129066.1 - 5 1742
BC035300.1 - 5 1816 4237864

View Full Product Details